KEY004181537 Key Organics / BIONET

2-[(2-carboxyethyl)sulfanyl]pyridine-3-carboxylic acid

  • MDL number MFCD02250406
  • Cas Number : 286472-02-4
  • Molecular Weight : 227.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 203 - 205
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/80238

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5R-0323_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart