KEY004181538 Key Organics / BIONET

2-amino-1-[(4-methoxyphenyl)methyl]-4,5,6,7-tetrahydro-1H-indole-3-carbonitrile

  • MDL number MFCD03425783
  • Cas Number : 113772-12-6
  • Molecular Weight : 281.359

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 130 - 132
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/80496

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5T-0002_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart