KEY004181540 Key Organics / BIONET

1-methyl-2-(2-phenylethyl)-3-(trifluoromethyl)-2,3-dihydro-1H-indol-3-ol

  • MDL number MFCD03425785
  • Cas Number : 439095-40-6
  • Molecular Weight : 321.343

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point Oil
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/80520

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5T-0047_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart