KEY004181541 Key Organics / BIONET

1-[(2-chloro-1,3-thiazol-5-yl)methyl]-1H-indole-3-carbaldehyde

  • MDL number MFCD03425786
  • Cas Number : 439095-43-9
  • Molecular Weight : 276.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 140 - 142
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/80522

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5T-0050_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart