KEY004181547 Key Organics / BIONET

5-cyano-4-methyl-3-phenylthiophene-2-carboxamide

  • MDL number MFCD03425789
  • Cas Number : 70541-99-0
  • Molecular Weight : 242.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 181 - 183
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/80600

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5T-0305_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart