KEY004181552 Key Organics / BIONET

3-methoxy-2-(prop-2-en-1-yloxy)benzaldehyde

  • MDL number MFCD03001271
  • Cas Number : 23343-06-8
  • Molecular Weight : 192.214

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point Oil
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/81909

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6G-945_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart