KEY004181556 Key Organics / BIONET

[3-(4-chlorophenyl)-1,2-oxazol-5-yl]methanamine

  • MDL number MFCD01935956
  • Cas Number : 66046-42-2
  • Molecular Weight : 208.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 113 - 115
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/82878

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6M-343S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart