KEY004181558 Key Organics / BIONET

2-[(carboxymethyl)sulfanyl]pyridine-3-carboxylic acid

  • MDL number MFCD02250402
  • Cas Number : 325704-15-2
  • Molecular Weight : 213.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 216 - 218
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/83632

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6R-0228_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart