KEY004181560 Key Organics / BIONET

(2E)-2-(2-methoxyphenyl)-3-phenylprop-2-enoic acid

  • MDL number MFCD03001273
  • Cas Number : 1657-65-4
  • Molecular Weight : 254.285

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 146 - 148
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/83659

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6R-0263_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart