KEY004181562 Key Organics / BIONET

(2E)-3-[4-(benzyloxy)phenyl]-2-phenylprop-2-enoic acid

  • MDL number MFCD03001274
  • Cas Number : 126582-18-1
  • Molecular Weight : 330.383

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 218 - 221
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/83707

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6R-0329_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart