KEY004181563 Key Organics / BIONET

4-phenyl-1H-pyrazolo[3,4-b]pyridin-3-amine

  • MDL number MFCD03001275
  • Cas Number : 174713-63-4
  • Molecular Weight : 210.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 262 - 264
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/83719

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6R-0346_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart