KEY004181564 Key Organics / BIONET

methyl 1-[(3,4-dichlorophenyl)methyl]piperidine-4-carboxylate

  • MDL number MFCD03425791
  • Cas Number : 383146-92-7
  • Molecular Weight : 302.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point Oil
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/83725

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6R-0353_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart