KEY004181566 Key Organics / BIONET

(2E)-3-[(4-acetylphenyl)carbamoyl]prop-2-enoic acid

  • MDL number MFCD00174964
  • Cas Number : 24870-12-0
  • Molecular Weight : 233.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 206 - 209
Iupac Name (2E)-4-(4-acetylanilino)-4-oxo-2-butenoic acid
Inchi Code 1S/C12H11NO4/c1-8(14)9-2-4-10(5-3-9)13-11(15)6-7-12(16)17/h2-7H,1H3,(H,13,15)(H,16,17)/b7-6+
Inchi Key CWSPWHGWDJSNIB-VOTSOKGWSA-N

Description

Melting Point 206 - 209
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/83943

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/6T-0030_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart