KEY004181568 Key Organics / BIONET

2,6-dihydroxy-5-{[4-(4-methoxyphenyl)piperazin-1-yl]methyl}-4-methylpyridine-3-carbonitrile

  • MDL number MFCD03425792
  • Cas Number : 439095-50-8
  • Molecular Weight : 354.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 193 - 195
Iupac Name 2,6-dihydroxy-5-{[4-(4-methoxyphenyl)-1-piperazinyl]methyl}-4-methylnicotinonitrile
Inchi Code 1S/C19H22N4O3/c1-13-16(11-20)18(24)21-19(25)17(13)12-22-7-9-23(10-8-22)14-3-5-15(26-2)6-4-14/h3-6H,7-10,12H2,1-2H3,(H2,21,24,25)
Inchi Key QLAZGSZNTGGSFZ-UHFFFAOYSA-N

Description

Melting Point 193 - 195
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/83957

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/6T-0063_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart