KEY004181569 Key Organics / BIONET

ethyl 1-(4-aminophenyl)piperidine-4-carboxylate

  • MDL number MFCD03425793
  • Cas Number : 439095-52-0
  • Molecular Weight : 248.326

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 40 - 42
Purity 95
Storage temp. Store under N2 at 4
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/83987

Safety Information

signalword Warning
pcodes P411,P422, P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6T-0212_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart