KEY004181570 Key Organics / BIONET

7-amino-3,4-dihydro-2H-1,4-benzoxazin-3-one

  • MDL number MFCD03425794
  • Cas Number : 105202-20-8
  • Molecular Weight : 164.164

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 215 - 218
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84003

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6T-0236_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart