KEY004181573 Key Organics / BIONET

2-(2-phenylethyl)-3-(trifluoromethyl)-1-azatricyclo[6.3.1.0^{4,12}]dodeca-4(12),5,7-trien-3-ol

  • MDL number MFCD03425797
  • Cas Number : 439095-60-0
  • Molecular Weight : 347.381

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 76 - 78
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84029

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6T-0279_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart