KEY004181574 Key Organics / BIONET

ethyl 4H-thieno[3,2-b]pyrrole-5-carboxylate

  • MDL number MFCD03425800
  • Cas Number : 46193-76-4
  • Molecular Weight : 195.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 131 - 132
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84048

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6T-0315_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart