KEY004181575 Key Organics / BIONET

ethyl 2-(benzylsulfanyl)-7-formyl-6-methylpyrazolo[3,2-b][1,3]thiazole-3-carboxylate

  • MDL number MFCD01837173
  • Cas Number : 263553-82-8
  • Molecular Weight : 360.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110 - 112
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84057

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6T-0331_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart