KEY004181582 Key Organics / BIONET

5-({[(4-chlorophenyl)methyl]sulfanyl}methyl)-4-(propan-2-yl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD01935945
  • Cas Number : 338982-45-9
  • Molecular Weight : 313.87

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 132 - 135
Iupac Name 5-{[(4-chlorobenzyl)sulfanyl]methyl}-4-isopropyl-4H-1,2,4-triazol-3-yl hydrosulfide
Inchi Code 1S/C13H16ClN3S2/c1-9(2)17-12(15-16-13(17)18)8-19-7-10-3-5-11(14)6-4-10/h3-6,9H,7-8H2,1-2H3,(H,16,18)
Inchi Key LPHBBWYJEHLAJI-UHFFFAOYSA-N

Description

Melting Point 132 - 135
Purity 95%
Country of Origin GB
Availability details for
Quantity
  • Availability
  • Add to Cart