KEY004181584 Key Organics / BIONET

2-(benzylsulfanyl)benzoic acid

  • MDL number MFCD00995231
  • Cas Number : 1531-80-2
  • Molecular Weight : 244.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 180 - 182
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/85986

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7K-659S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart