KEY004181588 Key Organics / BIONET

(2-chloro-1,3-thiazol-5-yl)methanol

  • MDL number MFCD03001277
  • Cas Number : 145015-15-2
  • Molecular Weight : 149.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 96@0.2mm Hg
Iupac Name (2-chloro-1,3-thiazol-5-yl)methanol
Inchi Code 1S/C4H4ClNOS/c5-4-6-1-3(2-7)8-4/h1,7H,2H2
Inchi Key PEOOQVDIOJQJHW-UHFFFAOYSA-N

Description

Melting Point 96@0.2mm Hg
Purity 97%
Storage temp. Store under N2 at 4
Physical Form Liquid,Low Melting Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/86708

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404,P411,P422
MSDS https://www.keyorganics.net/media/catalog/product/sds/7N-099_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart