KEY004181589 Key Organics / BIONET

1,1,1-trifluoro-3-[(2-phenylethyl)amino]propan-2-ol

  • MDL number MFCD03001278
  • Cas Number : 400878-20-8
  • Molecular Weight : 233.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 63 - 65
Iupac Name 1,1,1-trifluoro-3-[(2-phenylethyl)amino]-2-propanol
Inchi Code 1S/C11H14F3NO/c12-11(13,14)10(16)8-15-7-6-9-4-2-1-3-5-9/h1-5,10,15-16H,6-8H2
Inchi Key MLJRBCBVNZITOI-UHFFFAOYSA-N

Description

Melting Point 63 - 65
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/87044

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/7P-037_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart