KEY004181590 Key Organics / BIONET

1-[(3,4-dichlorophenyl)methyl]piperidine-4-carbohydrazide

  • MDL number MFCD03001279
  • Cas Number : 400878-29-7
  • Molecular Weight : 302.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 131 - 133
Iupac Name 1-(3,4-dichlorobenzyl)-4-piperidinecarbohydrazide
Inchi Code 1S/C13H17Cl2N3O/c14-11-2-1-9(7-12(11)15)8-18-5-3-10(4-6-18)13(19)17-16/h1-2,7,10H,3-6,8,16H2,(H,17,19)
Inchi Key ARYFFRRHNKOZFV-UHFFFAOYSA-N

Description

Melting Point 131 - 133
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/87293

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/7R-0214_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart