KEY004181592 Key Organics / BIONET

2,2,2-trichloro-1-[4-(4-fluorobenzoyl)-1H-pyrrol-2-yl]ethan-1-one

  • MDL number MFCD03001281
  • Cas Number : 453557-67-0
  • Molecular Weight : 334.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 202 - 204
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/87335

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7R-0269_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart