KEY004181593 Key Organics / BIONET

(3-methoxypropyl)urea

  • MDL number MFCD03425804
  • Cas Number : 1119-61-5
  • Molecular Weight : 132.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 76 - 78
Iupac Name N-(3-methoxypropyl)urea
Inchi Code 1S/C5H12N2O2/c1-9-4-2-3-7-5(6)8/h2-4H2,1H3,(H3,6,7,8)
Inchi Key SJCBTGYXKOFNHX-UHFFFAOYSA-N

Description

Melting Point 76 - 78
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/87337

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/7R-0274_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart