KEY004181603 Key Organics / BIONET

3-amino-3-(2-bromophenyl)propanoic acid

  • MDL number MFCD01871297
  • Cas Number : 117391-48-7
  • Molecular Weight : 244.088

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 221 - 223
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/87432

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7R-0619_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart