KEY004181605 Key Organics / BIONET

3-(4-bromophenyl)-3-{[(tert-butoxy)carbonyl]amino}propanoic acid

  • MDL number MFCD02093998
  • Cas Number : 282524-86-1
  • Molecular Weight : 344.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 195 - 196
Iupac Name 3-(4-bromophenyl)-N-(tert-butoxycarbonyl)-beta-alanine
Inchi Code 1S/C14H18BrNO4/c1-14(2,3)20-13(19)16-11(8-12(17)18)9-4-6-10(15)7-5-9/h4-7,11H,8H2,1-3H3,(H,16,19)(H,17,18)
Inchi Key ZAMLGGRVTAXBHI-UHFFFAOYSA-N

Description

Melting Point 195 - 196
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/87436

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/7R-0623_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart