KEY004181610 Key Organics / BIONET

3-{[(tert-butoxy)carbonyl]amino}-3-(3,4-dimethoxyphenyl)propanoic acid

  • MDL number MFCD01931739
  • Cas Number : 284492-37-1
  • Molecular Weight : 325.361

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 130 - 131
Purity 95
Storage temp. RT
Physical Form White to Off-white Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/87441

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/7R-0628_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart