KEY004181612 Key Organics / BIONET

3-{[(tert-butoxy)carbonyl]amino}-3-(3-fluorophenyl)propanoic acid

  • MDL number MFCD02090701
  • Cas Number : 284493-59-0
  • Molecular Weight : 283.299

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 120 - 121
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/87446

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7R-0633_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart