KEY004181621 Key Organics / BIONET

methyl 1-(4-chlorophenyl)-4-oxo-1,4-dihydropyridazine-3-carboxylate

  • MDL number MFCD03001289
  • Cas Number : 453557-76-1
  • Molecular Weight : 264.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 186 - 189
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/87637

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7R-1423_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart