KEY004181627 Key Organics / BIONET

ethyl 2-(4-chlorophenyl)-4-methyl-1,3-thiazole-5-carboxylate

  • MDL number MFCD00141958
  • Cas Number : 54001-12-6
  • Molecular Weight : 281.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 83 - 85
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/89239

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8J-003_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart