KEY004181629 Key Organics / BIONET

2-{[(4-methylphenyl)methyl]sulfanyl}benzoic acid

  • MDL number MFCD01935992
  • Cas Number : 104351-51-1
  • Molecular Weight : 258.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 183 - 187
Iupac Name 2-[(4-methylbenzyl)sulfanyl]benzoic acid
Inchi Code 1S/C15H14O2S/c1-11-6-8-12(9-7-11)10-18-14-5-3-2-4-13(14)15(16)17/h2-9H,10H2,1H3,(H,16,17)
Inchi Key KHKYMBMPMKQXCP-UHFFFAOYSA-N

Description

Melting Point 183 - 187
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/89498

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/8K-513S_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart