KEY004181630 Key Organics / BIONET

2-chloro-4-phenylpyridine-3-carbonitrile

  • MDL number MFCD01935958
  • Molecular Weight : 214.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 125 - 127.5
Iupac Name 2-chloro-4-phenylnicotinonitrile
Inchi Code 1S/C12H7ClN2/c13-12-11(8-14)10(6-7-15-12)9-4-2-1-3-5-9/h1-7H
Inchi Key GYTQTBOCDGGMRW-UHFFFAOYSA-N

Description

Melting Point 125 - 127.5
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/89840

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/8M-510S_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart