KEY004181636 Key Organics / BIONET

(5Z)-5-[(dimethylamino)methylidene]-3-ethyl-2-[(4H-1,2,4-triazol-4-yl)amino]-1,3-thiazolidin-4-one

  • MDL number MFCD05021865
  • Cas Number : 453557-84-1
  • Molecular Weight : 268.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 266.32
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/90470

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8R-0039_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart