KEY004181637 Key Organics / BIONET

2,2,2-trichloro-1-[4-(4-methoxybenzoyl)-1H-pyrrol-2-yl]ethan-1-one

  • MDL number MFCD03001290
  • Cas Number : 174417-17-5
  • Molecular Weight : 346.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 146 - 149
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/90499

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8R-0207_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart