KEY004181638 Key Organics / BIONET

methyl 4-(2,4-dichlorobenzoyl)-1H-pyrrole-2-carboxylate

  • MDL number MFCD03001291
  • Cas Number : 453557-85-2
  • Molecular Weight : 298.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 172 - 175
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/90556

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8R-0277_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart