KEY004181646 Key Organics / BIONET

2-oxo-4-(thiophen-2-yl)-1,2-dihydropyridine-3-carbonitrile

  • MDL number MFCD03001295
  • Cas Number : 174713-67-8
  • Molecular Weight : 202.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 291 - 293
Iupac Name 2-oxo-4-(2-thienyl)-1,2-dihydro-3-pyridinecarbonitrile
Inchi Code 1S/C10H6N2OS/c11-6-8-7(3-4-12-10(8)13)9-2-1-5-14-9/h1-5H,(H,12,13)
Inchi Key XCPYVKHGOXOHEJ-UHFFFAOYSA-N

Description

Melting Point 291 - 293
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/90594

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/8R-0329_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart