KEY004181647 Key Organics / BIONET

2-[1-(furan-2-yl)ethylidene]propanedinitrile

  • MDL number MFCD01570319
  • Cas Number : 62737-71-7
  • Molecular Weight : 158.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 86 - 89
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/90608

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8R-0348_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart