KEY004181650 Key Organics / BIONET

3-{[(tert-butoxy)carbonyl]amino}-3-(3-methoxyphenyl)propanoic acid

  • MDL number MFCD02090693
  • Cas Number : 284493-53-4
  • Molecular Weight : 295.335

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 77 - 79
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/90659

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8R-0613_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart