KEY004181651 Key Organics / BIONET

3-{[(tert-butoxy)carbonyl]amino}-3-(4-methylphenyl)propanoic acid

  • MDL number MFCD02090702
  • Cas Number : 284493-60-3
  • Molecular Weight : 279.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 168 - 170
Iupac Name N-(tert-butoxycarbonyl)-3-(4-methylphenyl)-beta-alanine
Inchi Code 1S/C15H21NO4/c1-10-5-7-11(8-6-10)12(9-13(17)18)16-14(19)20-15(2,3)4/h5-8,12H,9H2,1-4H3,(H,16,19)(H,17,18)
Inchi Key MBWMIEZHOLGJBM-UHFFFAOYSA-N

Description

Melting Point 168 - 170
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/90660

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/8R-0614_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart