KEY004181659 Key Organics / BIONET

3-amino-3-(3-hydroxyphenyl)propanoic acid

  • MDL number MFCD01871309
  • Cas Number : 102872-33-3
  • Molecular Weight : 181.191

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 222 - 223
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/90695

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8R-0664_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart