KEY004181661 Key Organics / BIONET

methyl 2-isothiocyanatothiophene-3-carboxylate

  • MDL number MFCD03001298
  • Cas Number : 126637-07-8
  • Molecular Weight : 199.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 59 - 61
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/90847

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8R-1327_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart