KEY004181662 Key Organics / BIONET

4-{[5-(trifluoromethyl)pyridin-2-yl]oxy}aniline

  • MDL number MFCD03001213
  • Cas Number : 71422-81-6
  • Molecular Weight : 254.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 44 - 47
Iupac Name 4-{[5-(trifluoromethyl)-2-pyridinyl]oxy}phenylamine
Inchi Code 1S/C12H9F3N2O/c13-12(14,15)8-1-6-11(17-7-8)18-10-4-2-9(16)3-5-10/h1-7H,16H2
Inchi Key LKVNUMLULPTKAU-UHFFFAOYSA-N

Description

Melting Point 44 - 47
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/91815

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/9F-067_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart