KEY004181663 Key Organics / BIONET

1-methyl-3-(trifluoromethyl)-5-[3-(trifluoromethyl)phenoxy]-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD01936007
  • Cas Number : 158712-29-9
  • Molecular Weight : 354.208

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 192 - 194
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/91818

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9F-301S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart