KEY004181664 Key Organics / BIONET

2-chloro-N-(2-methoxyphenyl)acetamide

  • MDL number MFCD00095394
  • Cas Number : 55860-22-5
  • Molecular Weight : 199.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp Oil
Iupac Name 2-chloro-N-(2-methoxyphenyl)acetamide
Inchi Code 1S/C9H10ClNO2/c1-13-8-5-3-2-4-7(8)11-9(12)6-10/h2-5H,6H2,1H3,(H,11,12)
Inchi Key IBXYYSDKRPMGKD-UHFFFAOYSA-N

Description

Melting Point Oil
Purity 95%
Physical Form Liquid,Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/92085

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/9G-909_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart