KEY004181667 Key Organics / BIONET

1-(4-methyl-1,2,5-oxadiazol-3-yl)ethan-1-one

  • MDL number MFCD00476459
  • Cas Number : 165067-10-7
  • Molecular Weight : 126.115

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 76@18mm Hg
Purity 95
Storage temp. RT
Physical Form Colourless to Yellow Liquid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/93041

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/9N-014_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart