KEY004181670 Key Organics / BIONET

4-(3-formyl-2,5-dimethyl-1H-pyrrol-1-yl)benzoic acid

  • MDL number MFCD00022463
  • Cas Number : 52034-38-5
  • Molecular Weight : 243.262

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 259 - 261
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/93419

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9P-735_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart