KEY004181672 Key Organics / BIONET

methyl 2-benzyl-2-cyano-3-phenylpropanoate

  • MDL number MFCD01314310
  • Cas Number : 42550-72-1
  • Molecular Weight : 279.339

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 57 - 60
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/93509

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9R-0037_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart