KEY004181676 Key Organics / BIONET

naphtho[2,1-b]furan-2-carbohydrazide

  • MDL number MFCD02571397
  • Cas Number : 53524-88-2
  • Molecular Weight : 226.235

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 252 - 255
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/93573

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9R-0261_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart